Learn More
Description
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq
Specifications
Specifications
| Antigen | FABP4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4D12 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant FABP4. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FABP4 |
| Gene Accession No. | BC003672 |
| Gene Alias | A-FABP/aP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.