Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512352
Description
This antibody is predicted to react with bovine, porcine and rat based on sequence homology.
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
Specifications
FABP4 | |
Polyclonal | |
FABP4 | |
Fatty acid-binding protein, adipocyte; A-FABP; AFABP; Fatty acid-binding protein 4; Adipocyte lipid-binding protein; ALBP; FABP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11770, 2167 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P15090, P04117 | |
FABP4 | |
KLH conjugated synthetic peptide between 1-30 amino acids from human FABP4 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction