Learn More
fatty acid binding protein 5 (psoriasis-associated), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FABP5 protein.
Supplier: Abnova Corporation H00002171B02
Description
This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated in psoriasis tissue. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. The human genome contains many pseudogenes similar to this locus. [provided by RefSeq
Sequence: MATVQQLEGRWRLVDSKGFDEYMKELGVGIALRKMGAMAKPDCIITCDGKNLTIKTESTLKTTQFSCTLGEKFEETTADGRKTQTVCNFTDGALVQHQEWDGKESTITRKLKDGKLVVECVMNNVTCTRIYEKVESpecifications
| fatty acid binding protein 5 (psoriasis-associated) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FABP5 protein. | |
| FABP5 | |
| E-FABP/EFABP/PA-FABP/PAFABP | |
| Mouse | |
| 50 μL | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_001444 | |
| FABP5 | |
| FABP5 (NP_001435.1, 1 a.a. ∼ 135 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 2171 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.