Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP5/E-FABP Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB3077
This item is not returnable.
View return policy
Description
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.Specifications
FABP5/E-FABP | |
Monoclonal | |
LYOPH | |
Western Blot 1 μg/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 μg/mL, CyTOF-ready, Knockout Validated | |
Q01469 | |
FABP5 | |
E. coli-derived recombinant human FABP5/E-FABP Ala2-Glu135 Accession # Q01469 | |
100 μg | |
Primary | |
Detects human FABP5/E-FABP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 6, 7, or 9 is observed. | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Immunoassay | |
311215 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EFABP, E-FABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2171 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction