Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Factor B, Polyclonal

Catalog No. 3412722ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
34-127-22ML 2 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-127-22ML Supplier MilliporeSigma™ Supplier No. 3412722ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects Factor B Clone: in Human samples, and it is validated for Immunodiffusion, Immunoelectrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor B
Applications Immunodiffusion, Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Host Species Goat
Immunogen purified, human factor B
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Factor B.
Target Species Human
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.