Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Mouse anti-Human, MilliporeSigma™

Catalog No. MAB038MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB038MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB038MI Supplier MilliporeSigma Supplier No. MAB038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Factor VIII Antibody, 83 kDa light chain is an antibody against Factor VIII for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor VIII
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Liquid. Contains no preservative.
Gene Accession No. NM_000132.2; NM_019863.2
Gene Symbols F8, OTTHUMP00000061446, F8C, HEMA, AHF, DXS1253E, FVIII, F8B
Host Species Mouse
Immunogen Purified Human Factor VIII.
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.