Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 06711MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-711-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-711-MI Supplier MilliporeSigma Supplier No. 06711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect FAS-associated death domain protein using this rabbit polyclonal Anti-FADD, Cat. No. 06-711. It has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADD
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity purified rabbit Ig in a solution containing 0.1M Tris-glycine, pH 7.0 and 0.05% sodium azide
Gene Symbols FADD; GIG3; MGC8528; MORT1
Host Species Rabbit
Immunogen His-tagged full-length fusion protein corresponding to human FADD
Purification Method Affinity Purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003824
Target Species Human
Content And Storage −20°C for 24 months. Aliquot to avoid repeated freezing and thawing
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.