Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Mouse anti-Human, Mouse, Clone: 1F7, MilliporeSigma™

Catalog No. 05486MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-486-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-486-MI Supplier MilliporeSigma Supplier No. 05486
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FADD Antibody, clone 1F7 detects level of FADD and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADD
Applications Western Blot
Classification Monoclonal
Clone 1F7
Conjugate Unconjugated
Formulation PBS, pH 7.2 and 50% glycerol
Gene Accession No. NM_003824
Gene Symbols FADD, GIG3, MGC8528, MORT1
Host Species Mouse
Immunogen GST-tagged full-length fusion protein corresponding to human FADD
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.