Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAIM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520312
Description
A suggested positive control is human spleen tissue lysate. PA5-20312 can be used with blocking peptide PEP-0432.
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
Specifications
FAIM | |
Polyclonal | |
Unconjugated | |
FAIM | |
Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23873, 55179 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9NVQ4, Q9WUD8 | |
FAIM | |
A 14 amino acid peptide from near the carboxy terminus of human FAIM. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction