Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAK Mouse, Unlabeled, Clone: 77/FAK, BD

Catalog No. BDB610088 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610088 150 μg
BDB610087 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610088 Supplier BD Biosciences Supplier No. 610088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that colocalizes with integrins in focal adhesions. This cellular localization is directed by a 125 amino acid sequence at the C-terminus called the “Focal Adhesion Targeting sequence” (FAT). The binding of extracellular matrix ligands to integrins triggers autophosphorylation and activation of FAK. This creates binding sites for SH2 domains of intracellular signaling molecules such as src, PI3 kinase, and Grb2. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions has led to substantial speculation about its function. Although FAK's precise role has not been elucidated, proposed possibilities include regulating cell motility, cell growth, cytoskeletal organization, and adhesion-dependent cell survival.

Host Species: Mouse
Clone: 77
Isotype: IgG1
Species Reactivity: Human
Immunogen: Chicken FAK aa. 354-533
Formula Weight [Chemical]: 116-125kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAK
Applications Western Blot
Classification Monoclonal
Clone 77/FAK
Concentration 250μg/mL
Conjugate Unconjugated
Description Focal Adhesion Kinase
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Chicken FAK aa. 354-533
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.