Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAK Rabbit anti-Human, Mouse, Rat, Hamster, Polyclonal, MilliporeSigma™

Catalog No. 06543MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-543-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-543-MI Supplier MilliporeSigma Supplier No. 06543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-FAK Antibody is validated for use in western blot, immunoprecipitation, IC for the detection of FAK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAK
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide). Store at −20°C.
Gene Symbols PTK2; pp125FAK; FADK; FAK1; FAK
Host Species Rabbit
Immunogen pGEX-derived fusion protein containing residues 748-1053 of human FAK
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005607 / NM_153831
Target Species Hamster, Human, Mouse, Rat
Content And Storage Stable for 1 year at .
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.