Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAK (pY397) (Focal Adhesion Kinase (pY397)) Mouse anti-Human, Unlabeled, Clone: 18, BD

Catalog No. BDB611806 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611806 50 μg
BDB611807 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611806 Supplier BD Biosciences Supplier No. 611806
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that colocalizes with integrins in focal adhesions. This cellular localization is directed by a 125 amino acid sequence at the C-terminus called “Focal Adhesion Targeting” sequence (FAT). The binding of extracellular matrix ligands to integrins triggers autophosphorylation at Tyr-397, and activation of FAK through phosphorylation of Tyr residues (Tyr-576 and Tyr-577) in the kinase domain activation loop. For example, cell adhesion to a fibronectin substratum involves concurrent activation of Src and phosphorylation of the FAK activation loop. In addition, phosphorylation of other Tyr residues (Tyr-925 and Tyr-861) creates binding sites for SH2 domains of intracellular signaling molecules such as Src, PI3 kinase, and Grb2. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions is important for FAK activation level and for FAK interaction with a variety of substrates localized to sites of cell adhesion. Thus, FAK activity is regulated by a complex set of phosphorylation sites, and this phospho-regulation could be important for cell motility, cell growth, cytoskeletal organization and adhesion-dependent cell survival.

Host Species: Mouse
Clone: 18
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human FAK (pY397)

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAK (pY397) (Focal Adhesion Kinase (pY397))
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Description Focal Adhesion Kinase (pY397)
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human FAK (pY397)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.