Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM120A Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABE792MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE792MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE792MI Supplier MilliporeSigma Supplier No. ABE792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-FAM120A Antibody is an antibody against FAM120A for use in western blotting, immunohistochemistry and immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM120A
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols FAM120A; C9orf10; KIAA0183; OSSA
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the N-terminus of human FAM120A.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.