Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM48A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528049
Description
Recommended positive controls: 293T. Predicted reactivity: Rat (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Required for MAP kinase p38 (MAPK11, MAPK12, MAPK13 and/or MAPK14) activation during gastrulation. Required for down-regulation of E-cadherin during gastrulation by regulating E-cadherin protein level downstream from NCK-interacting kinase (NIK) and independently of the regulation of transcription by FGF signaling and Snail. Required for starvation-induced ATG9A trafficking during autophagy. [UniProt].
Specifications
FAM48A | |
Polyclonal | |
Unconjugated | |
SUPT20H | |
bA421P11.4; C13; C13orf19; Fam48a; family with sequence similarity 48, member A; FP757; p38-interacting protein; P38IP; protein FAM48A; RGD1307812; SPT20; SPT20 homolog, SAGA complex component; suppressor of Ty 20; suppressor of Ty 20 homolog; Supt20; SUPT20H; transcription factor (p38 interacting protein); transcription factor SPT20 homolog; tumor rejection antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55578 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NEM7 | |
SUPT20H | |
Recombinant fragment corresponding to a region within amino acids 151 and 440 of Human FAM48A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction