Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM50A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535258
Description
This antibody is predicted to react with mouse and Xenopus and zebrafish based on sequence homology.
May be a DNA-binding protein or transcriptional factor.
Specifications
| FAM50A | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q14320 | |
| FAM50A | |
| KLH conjugated synthetic peptide between 311-339 amino acids from the C-terminal region of human FAM50A | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| FAM50A | |
| FAM50A;DXS9928E; HXC26; XAP5; Protein FAM50A; Protein FAM50A; Protein HXC-26; Protein FAM50A; Protein XAP-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9130 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction