Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM58A, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89208507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-208-507 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-208-507 Supplier Abnova Corporation Supplier No. H00092002K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human FAM58A peptide using ARM Technology.

Mutations in this gene have been shown to cause an X-linked dominant STAR syndrome that typically manifests syndactyly, telecanthus and anogenital and renal malformations. The protein encoded by this gene contains a cyclin-box-fold domain which suggests it may have a role in controlling nuclear cell division cycles. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen FAM58A
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description family with sequence similarity 58, member A
Formulation In 1X PBS, pH 7.4
Gene FAM58A
Gene Accession No. FAM58A
Gene Alias MGC29729, STAR
Gene Symbols FAM58A
Host Species Rabbit
Immunogen A synthetic peptide of human FAM58A
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92002
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.