Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM5C Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526496
Description
This antibody is predicted to react with mouse and rat based on sequence homology.

Specifications
FAM5C | |
Polyclonal | |
BRINP3 | |
BRINP3, DBCCR1L, DBCCR1L1, RP11-445K1.1, DBCCR1-like protein 1, protein FAM5C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339479 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q76B58 | |
BRINP3 | |
KLH conjugated synthetic peptide between 40-68 amino acids from the N-terminal region of human FAM5C | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction