Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCA Mouse anti-Human, Clone: 5G9, MilliporeSigma™

Catalog No. MABC557MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC557MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC557MI Supplier MilliporeSigma Supplier No. MABC557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FANCA Antibody, clone 5G9 is a highly specific mouse monoclonal antibody, that targets FANCA and has been tested in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCA
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5G9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000126
Gene Symbols FANCA, FAA, FACA, FANCH
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human FANCA.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.