Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCB, Mouse, Clone: 2B10, Abnova™

Catalog No. 89022050 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-050 Supplier Abnova Corporation Supplier No. H00002187M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FANCB.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group B. Alternative splicing results in two transcript variants encoding the same protein. [provided by RefSeq

Sequence: GSENFLIDNMAFTLEKELVTLSSLSSAIAKHESNFMQRCEVSKGKSSVVAAALSDRRENIHPYRKELQREKKKMLQTNLKVSGALYREITLKVAEVQLKSDFAAQKLSN

Specifications

Antigen FANCB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FANCB.
Formulation PBS with no preservative; pH 7.4
Gene FANCB
Gene Accession No. NM_152633
Gene Alias FA2/FAAP90/FAAP95/FAB/FACB
Gene Symbols FANCB
Host Species Mouse
Immunogen FANCB (NP_689846.1, 750 a.a. ∼ 858 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2187
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.