Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCC Mouse anti-Rat, Mouse, Human, Clone: 8F3, MilliporeSigma™

Catalog No. MABC524MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC524MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC524MI Supplier MilliporeSigma Supplier No. MABC524
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-FANCC Antibody, clone 8F3 is validated for use in Western Blotting and Immunoprecipitation and Immunocytochemistry for the detection of FANCC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCC
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8F3
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000127
Gene Symbols FANCC, FAC, FACC
Host Species Mouse
Immunogen Linear peptide corresponding to Human FANCC.
Purification Method Protein G purified
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.