Learn More
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group G. [provided by RefSeq
Specifications
Specifications
| Antigen | FANCG |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot |
| Classification | Monoclonal |
| Clone | 2C8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant FANCG. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FANCG |
| Gene Accession No. | BC011623 |
| Gene Alias | FAG/XRCC9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.