Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fanconi anemia, complementation group G, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017623 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-623 Supplier Abnova Corporation Supplier No. H00002189D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human FANCG protein.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group G. [provided by RefSeq

Sequence: MSRQTTSVGSSCLDLWREKNDRLVRQAKVAQNSGLTLRRQQLAQDALEGLRGLLHSLQGLPAAVPVLPLELTVTCNFIILRASLAQGFTEDQAQDIQRSLERVLETQEQQGPRLEQGLRELWDSVLRASCLLPELLSALHRLVGLQAALWLSADRLGDLALLLETLNGSQSGASKDLLLLLKTWSPPAEELDAPLTLQDAQGLKDVLLTAFAYRQGLQELITGNPDKALSSLHEAASGLCPRPVLVQVYTALGSCHRKMGNPQRALLYLVAALKEGSAWGPPLLEASRLYQQLGDTTAELESLELLVEALNVPCSSKAPQFLIEVELLLPPPDLASPLHCGTQSQTKHILASRCLQTGRAGDAAEHYLDLLALLLDSSEPRFSPPPSPPGPCMPEVFLEAAVALIQAGRAQDALTLCEELLSRTSSLLPKMSRLWEDARKGTKELPYCPLWVSATHLLQGQAWVQLGAQKVAISEFSRCLELLFRATPEEKEQGAAFNCEQGCKSDAALQQLRAAALISRGLEWVASGQDTKALQDFLLSVQMCPGNRDTYFHLLQTLKRLDRRDEATALWWRLEAQTKGSHEDALWSLPLYLESYLSWIRPSDRDAFLEEFRTSLPKSCDL

Specifications

Antigen Fanconi anemia, complementation group G
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human FANCG protein.
Formulation PBS with no preservative; pH 7.4
Gene FANCG
Gene Accession No. NM_004629
Gene Alias FAG/XRCC9
Gene Symbols FANCG
Host Species Rabbit
Immunogen FANCG (NP_004620.1, 1 a.a. ∼ 622 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2189
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.