Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FAP-1/FAS-Associated Phosphatase, Polyclonal, Epredia™ LabVision

Catalog No. RB10457P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RB10457P0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB10457P0 Supplier Epredia Supplier No. RB10457P0

Rabbit Polyclonal Antibody

A novel protein tyrosine phosphatase, Fap-1 (for FAS-associated phosphatase, originally designated PTP-BAS), has been shown to associated with the carboxy terminus fifteen amino acids of FAS. Three isoforms of the protein result from alternative RNA splicings, the longest of which encodes a protein 2485 amino aicds in lenghth. Although lacking a transmembrane region, Fap-1 does contain a membrane-binding domain, similar to that found in cytoskeleton-associated proteins such as ezrin. Fap-1 does not seem to associate with CD40 or death domain proteins such as TNF-RI and TNF-RII.

Host Species: Rabbit
Cross Reactivity: Human. Others not tested.
Epitope: aa 2290-2491
Immunogen: Recombinant protein encoding aa 2290-2491 of human FAP-1.
Molecular Weight: 280kDa
Positive Control: Placenta
Cellular Localization: Cytoplasmic

Immunohistochemistry (Formalin/Paraffin)

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen FAP-1/FAS-Associated Phosphatase
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding aa 2290-2491 of human FAP-1.
Purification Method Placenta
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2–8 degrees
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.