Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-FAP Clone: F11-24,

Catalog No. OP188100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP188100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP188100UG Supplier MilliporeSigma™ Supplier No. OP188100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FAP Clone: F11-24 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation

FAP is a cell surface glycoprotein of reactive human fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAP
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone F11-24
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen HEK293 cells transfected with an expression vector encoding full-length human FAP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes fibroblast activation protein (FAP).
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.