Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Farnesyl Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB4073MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB4073MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB4073MI Supplier MilliporeSigma Supplier No. AB4073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Farnesyl Antibody detects level of Farnesyl and has been published and validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Farnesyl
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Sterile liquid in phosphate buffered saline and 0.08% sodium azide.
Host Species Rabbit
Immunogen Antibody developed using farnesyl cysteine conjugated to KLH.
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.