Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas Rat anti-Bovine, Mouse, Clone: 7C10, MilliporeSigma™

Catalog No. 05351MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-351-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-351-MI Supplier MilliporeSigma Supplier No. 05351
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Fas Antibody, clone 7C10 is an antibody against Fas for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fas
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7C10
Conjugate Unconjugated
Formulation 0.1MTris-glycine, 0.15MNaCl, pH 7.4 with 0.05% sodium azide
Gene Accession No. NM_152877
Gene Symbols FAS, OTTHUMP00000059646, LFG, FASTM, ALPS1A, TNFRSF6, CD95, FAS1, FAIM2, APT1, NMP35, APO-1
Host Species Rat
Immunogen recombinant protein containing the cytoplasmic domain of mouse Fas
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.