Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas Mouse anti-Human, Clone: CH11, MilliporeSigma™

Catalog No. 05201MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-201-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-201-MI Supplier MilliporeSigma Supplier No. 05201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Fas using this Anti-Fas Antibody (human, activating), clone CH11 has been published and validated for use in Flow Cytometry (FC), Immunocytochemistry (IC) and Western Blot (WB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fas
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone CH11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM in buffer containing PBS, pH 7.2, with 50% glycerol. Liquid at −20deg;C.
Gene Accession No. NM_152877
Gene Symbols FAS, OTTHUMP00000059646, LFG, FASTM, ALPS1A, TNFRSF6, CD95, FAS1, FAIM2, APT1, NMP35, APO-1
Host Species Mouse
Immunogen FS-7 (human diploid fibroblast cell line). Clone CH-11.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.