Learn More
Fas Ligand Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Fas Ligand |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Peptide affinity purified immunoglobulin. Liquid in PBS, containing 0.1% BSA. |
| Gene Symbols | FAS; CD95; FAS1; FAIM2; OTTHUMP00000059646; LFG; APT1; FASTM; NMP35; APO-1; ALPS1A; TNFRSF6 |
| Host Species | Rabbit |
| Immunogen | A 13 AA peptide sequence mapping near the C-terminus of human FAS ligand (Mita et al. 1994). This peptide is predicted to be extracellular. The peptide was synthesized with C-terminal Cysteine, coupled to KLH. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.