Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas Ligand Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1665
Description
Detect Fas Ligand using this Anti-Fas Ligand Antibody validated for use in ELISA and western blotting.
Specifications
Fas Ligand | |
Polyclonal | |
Liquid antiserum containing 0.05% sodium merthiolate as a preservative. | |
Rabbit | |
100 μL | |
NM_000639.1 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
ELISA, Western Blot | |
Unconjugated | |
FASLG; FASL; CD95L; CD178; TNFSF6; FasL; APT1LG1; APTL | |
An 18 amino acid peptide sequence (aa 130-146) from rat FASL. This sequence is known to be 94% homologous with mouse FASL, differing by one amino acid. This peptide region does not show significant homology with human FASL, with only four amin | |
Primary | |
Mouse, Rat | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction