Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin Mouse anti-Human, Mammals, Echinoderms, Gerbil, Sea Urchin, Clone: 55K2, MilliporeSigma™

Catalog No. MAB3582MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3582MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3582MI Supplier MilliporeSigma Supplier No. MAB3582
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Fascin Antibody, clone 55K2 is validated for use in western blot, immunocytochemistry, immunohistochemistry (paraffin) for the detection of Fascin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fascin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 55K2
Conjugate Unconjugated
Formulation Liquid in 0.02M Phosphate buffer with 0.25M NaCl and 0.1% sodium azide.
Gene Accession No. NM_003088.2
Gene Symbols FSCN1, FAN1, HSN, FLJ38511, SNL, Fascin, p55
Host Species Mouse
Immunogen Human fascin
Quantity 100 μg
Primary or Secondary Primary
Target Species Echinodermata, Gerbil, Human, Sea urchin
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.