Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-FASN Clone: 3F2-1F3,

Catalog No. ST154950UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST154950UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST154950UG Supplier MilliporeSigma™ Supplier No. ST154950UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FASN Clone: 3F2-1F3 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting

FASN is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-α), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-α.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASN
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3F2-1F3
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant human FASN (aa 1-440) expressed as GST fusion protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the FASN protein in 293 and HeLa cells and human breast carcinoma tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.