Learn More
Description
Includes
Rabbit affinity purified polyclonal anti-FASN (100μg); Mouse monoclonal anti-FASN, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The enzyme encoded by this gene is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-α (ER-α), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-α.
ELISA
Specifications
Specifications
| Antigen | FASN |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FASN. |
| Gene | FASN |
| Gene Alias | FAS/MGC14367/MGC15706/OA-519/SDR27X1 |
| Gene Symbols | FASN |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.