Learn More
Fatty Acid Amide Hydrolase Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Fatty Acid Amide Hydrolase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 100mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide. |
| Host Species | Rabbit |
| Immunogen | A 17 amino acid peptide sequence near the N-terminus of human FAAH (Patricelli et al. 1998; Maccarone et al. 1998; Cravatt et al. 1996; Giang et al. 1997; Thomas et al. 1997; Wan et al. 1998). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.