Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fatty-acid binding protein 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS312
Description
Detect Fatty Acid Binding Protein using this rabbit polyclonal antibody, Anti-Fatty-acid binding protein 5 Antibody validated for use in western blotting.
Specifications
Fatty-acid binding protein 5 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide. | |
Rabbit | |
Protein G purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
FABP5 | |
Recombinant protein corresponding to human Fatty-acid binding protein 5. | |
100 μg | |
NP_001435 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction