Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fatty Acid Synthase Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB610963 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610963 150 μg
BDB610962 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610963 Supplier BD Biosciences Supplier No. 610963
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fatty acid biosynthesis occurs in all living organisms and provides essential components of biological membranes as well as a form of energy storage. Animal fatty acid synthase (FAS) is a multifunctional enzyme that catalyzes the synthesis of long-chain fatty acids via sequential condensation of two-carbon units from malonyl-CoA, an intermediate derived from the carboxylation of Acetyl-CoA. FAS is a homodimer of a multifunctional subunit protein that contains seven distinct activities and a site for the prosthetic group 4'-phosphopantetheine (acyl carrier protein). These domains are oriented from N-terminus to C-terminus as follows: β-keto-acyl synthase, acetyl and malonyl transacylases, enoyl reductase, ketoacyl reductase, acyl carrier protein, and thioesterase. Although all domains are found on each subunit, they are only active following the homodimerization of subunits in an antiparallel (head-to-tail) orientation. This juxtaposition and cooperation between domains forms two centers for acyl chain assembly. Alternative substrates and chain-terminating mechanisms allow for the production of a variety of fatty acids with different lengths and structures.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fatty Acid Synthase
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Fatty Acid Synthase aa. 9-202
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rabbit, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.