Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAU, Mouse, Clone: 3C10, Abnova™

Catalog No. 89022061 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-061 Supplier Abnova Corporation Supplier No. H00002197M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FAU.

This gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). It encodes a fusion protein consisting of the ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. It has been proposed that the fusion protein is post-translationally processed to generate free fubi and free ribosomal protein S30. Fubi is a member of the ubiquitin family, and ribosomal protein S30 belongs to the S30E family of ribosomal proteins. Whereas the function of fubi is currently unknown, ribosomal protein S30 is a component of the 40S subunit of the cytoplasmic ribosome. Pseudogenes derived from this gene are present in the genome. Similar to ribosomal protein S30, ribosomal proteins S27a and L40 are synthesized as fusion proteins with ubiquitin. [provided by RefSeq

Sequence: MQLFVRAQELHTFEVTGQETVAQIKAHVASLEGIAPEDQVVLLAGAPLEDEATLGQCGVEALTTQEVAGRMLGGKVHGSLARAGKVRGQTPKVAKQEKKKKKTGRAKRRMQYNRRFVNVVPTFGKKKGPNANS

Specifications

Antigen FAU
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant FAU.
Formulation PBS with no preservative; pH 7.4
Gene FAU
Gene Accession No. BC033877
Gene Alias FAU1/FLJ22986/Fub1/Fubi/MNSFbeta/RPS30
Gene Symbols FAU
Host Species Mouse
Immunogen FAU (AAH33877, 1 a.a. ∼ 133 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2197
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.