Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box and leucine-rich repeat protein 17, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004712 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-712 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-712 Supplier Abnova Corporation Supplier No. H00064839A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FBXL17.

Members of the F-box protein family, such as FBXL17, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: QNLKELYLVSCKITDYALIAIGRYSMTIETVDVGWCKEITDQGATLIAQSSKSLRYLGLMRCDKVNEVTVEQLVQQYPHITFSTVLQDCKRTLERAYQMGWTPNMSAASS

Specifications

Antigen F-box and leucine-rich repeat protein 17
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXL17.
Formulation 50% glycerol
Gene FBXL17
Gene Accession No. NM_022824
Gene Alias DKFZp434C1715/FBXO13/Fbl17/Fbx13/MGC161422/MGC161424
Gene Symbols FBXL17
Host Species Mouse
Immunogen FBXL17 (NP_073735, 194 a.a. ∼ 303 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64839
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.