Learn More
F-box and leucine-rich repeat protein 18, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human FBXL18 protein.
Supplier: Abnova Corporation H00080028D01P
Description
Members of the F-box protein family, such as FBXL18, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MHAVPRGFGKKVRVGVQSCPPLLGPGVPPALLRVLVSAEEPALPGTPRADWVQLLLRHAPQRARHPQLAPTLQPRTECRGLGGGRHRPAGLPAAPDARTAAQRPYGLRAGQYRPAVPAVAVPVAGQPGHDGEGGVHARALRHVEALQAAEGPQAGAALLQRQRPVLPGAEPVPLAAAPVPGLSQRHPPARCRAGLHGSLPAGCHVPPVHRGVPRHLQEPAAVASPQLPGRAARVKRRHLPSAPRGPDRRHPGRPPGAPGSpecifications
| F-box and leucine-rich repeat protein 18 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human FBXL18 protein. | |
| FBXL18 | |
| FLJ10776/FLJ11467/FLJ26934/FLJ38075/FLJ41541/Fbl18 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC013435.1 | |
| FBXL18 | |
| FBXL18 (AAH13435.1, 1 a.a. ∼ 259 a.a) full-length human protein. | |
| 100 μg | |
| Ubiquitin/Proteasome | |
| 80028 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.