Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box and leucine-rich repeat protein 5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-586 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-586 Supplier Abnova Corporation Supplier No. H00026234A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FBXL5.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]

Sequence: LIYFGSEKSDQETGRVLLFLSLSGCYQITDHGLRVLTLGGGLPYLEHLNLSGCLTITGAGLQDLVSACPSLNDEYFYYCDNINGPHADTASGCQNLQCGFRACCRSGE

Specifications

Antigen F-box and leucine-rich repeat protein 5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXL5.
Formulation 50% glycerol
Gene FBXL5
Gene Accession No. NM_012161
Gene Alias FBL4/FBL5/FLR1
Gene Symbols FBXL5
Host Species Mouse
Immunogen FBXL5 (NP_036293, 584 a.a. ∼ 691 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 26234
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.