Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 30 (A01), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89004738 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-738 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-738 Supplier Abnova Corporation Supplier No. H00084085A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FBXO30.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it is upregulated in nasopharyngeal carcinoma. [provided by RefSeq]

Sequence: MVILQWGKRKYPEGNSSWQIKEKVWRFSTAFCSVNEWKFADILSMADHLKKCSYNVVEKREEAIPLPCMCVTRELTKEGRSLRSVLKPVL

Specifications

Antigen F-box protein 30
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXO30.
Formulation 50% glycerol
Gene FBXO30
Gene Accession No. NM_032145
Gene Alias FLJ41030/Fbx30/MGC21674
Gene Symbols FBXO30
Host Species Mouse
Immunogen FBXO30 (NP_115521, 656 a.a. ∼ 745 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84085
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.