Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 34, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007787 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-787 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-787 Supplier Abnova Corporation Supplier No. H00055030B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FBXO34 protein.

Members of the F-box protein family, such as FBXO34, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MHLKPYWKLQKKEHPPEVSRETQRTPMNHQKAVNDETCKASHITSSVFPSASLGKASSRKPFGILSPNVLCSMSGKSPVESSLNVKTKKNAPSATIHQGEEEGPLDIWAVVKPGNTKEKIAFFASHQCSNRIGSMKIKSSWDIDGRATKRRKKSGDLKKAKVQVERMREVNSRCYQPEPFACGIEHCSVHYVSDSGDGVYAGRPLSVIQMVAFLEQRASALLASCSKNCTNSPAIVRFSGQSRGVPAVSESYSAPGACEEPTERGNLEVGEPQSEPVRVLDMVAKLESECLKRQGQREPGSLSRNNSFRRNVGRVLLANSTQADEGKTKKGVLEAPDTQVNPVGSVSVDCGPSRADRCSPKEDQAWDGASQDCPPLPAGVSFHIDSAELEPGSQTAVKNSNRYDVEMTDELVGLPFSSHTYSQASELPTDAVDCMSRELVSLTSRNPDQRKESLCISITVSKVDKDQPSILNSCEDPVPGMLFFLPPGQHLSDYSQLNESTTKESSEASQLEDAAGGDSASEEKSGSAEPFVLPASSVESTLPVLEASSWKKQVSHDFLETRFKIQQLLEPQQYMAFLPHHIMVKIFRLLPTKSLVALKCTCCYFKFIIEYYNIRPADSRWVRDPRYREDPCKQCKKKYVKGDVSLCRWHPKPYCQALPYGPGYWMCCHRSQKGFPGCKLGLHDNHWVPACHSFNRAIHKKAKGTEAEEEY

Specifications

Antigen F-box protein 34
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FBXO34 protein.
Formulation No additive
Gene FBXO34
Gene Accession No. NM_017943.2
Gene Alias CGI-301/DKFZp547C162/FLJ20725/Fbx34/MGC126434/MGC126435
Gene Symbols FBXO34
Host Species Mouse
Immunogen FBXO34 (NP_060413.2, 1 a.a. ∼ 711 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55030
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.