Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 40, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018490 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-490 Supplier Abnova Corporation Supplier No. H00051725D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human FBXO40 protein.

Members of the F-box protein family, such as FBXO40, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MGKARRSPPGHHRHCEGCFNRHCHIPVEPNTSCLVISCHLLCGATFHMCKEAEHQLLCPLEQVPCLNSEYGCPLSMSRHKLAKHLQVCPASVVCCSMEWNRWPNVDSETTLHENIMKETPSEECLDTALALQDQKVLFRSLKMVELFPETREATEEEPTMNGETSVEEMGGAVGGVDIGLVPHGLSATNGEMAELSQEEREVLAKTKEGMDLVKFGQWENIFSKEHAASALTNSSASCESKNKNDSEKEQISSGHNMVEGEGAPKKKEPQENQKQQDVRTAMETTGLAPWQDGVLERLKTAVDAKDYNMYLVHNGRMLIHFGQMPACTPKERDFVYGKLEAQEVKTVYTFKVPVSYCGKRARLGDAMLSCKPSEHKAVDTSDLGITVEDLPKSDLIKTTLQCALERELKGHVISESRSIDGLFMDFATQTYNFEPEQFSSGTVLADLTAATPGGLHVELHSECVTRRHNKSSSAFTFTCNKFFRRDEFPLHFKNVHTDIQSCLNGWFQHRCPLAYLGCTFVQNHFRPPGQKAKVIYSQELKTFAIKPEVAPELSEGRKNNHLLGHGGKSQNSLTSLPLEILKYIAGFLDSVSLAQLSQVSVLMRNICATLLQERGMVLLQWKKKRYSHGGTSWRVHREIWQFSSLFSKIKSWEFNEVTSMSEHLKSCPFNIVEHKTDPILLTSMCQPREQARESLVSTFRIRPRGRYVS

Specifications

Antigen F-box protein 40
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human FBXO40 protein.
Formulation PBS with no preservative; pH 7.4
Gene FBXO40
Gene Accession No. NM_016298.1
Gene Alias FBX40/KIAA1195/MGC129902/MGC129903
Gene Symbols FBXO40
Host Species Rabbit
Immunogen FBXO40 (NP_057382.1, 1 a.a. ∼ 709 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51725
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.