Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box and WD repeat domain containing 11, Mouse, Clone: 2E7, Abnova™

Catalog No. 89014283 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-283 Supplier Abnova Corporation Supplier No. H00023291M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FBXW11.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class and, in addition to an F-box, contains multiple WD40 repeats. This gene contains at least 14 exons, and its alternative splicing generates 3 transcript variants diverging at the presence/absence of two alternate exons. [provided by RefSeq]

Sequence: MEPDSVIEDKTIELMNTSVMEDQNEDESPKKNTLWQISNGTSSVIVSRKRPSEGNYQKEKDLCIKYFDQWSESDQVEFVEHLISRMCHYQHGHINSYLKPMLQRDFITALPEQGLDHIAENILSYLDARSLCAAELVCKEWQRVISEGMLWKKLIERMVRTDPLWKGLSERRGWDQYLFKNRPTDGPPNSFYRSLYPKIIQDIETIESNWRCGRHNLQRIQCRSENSKGVYCLQYDDEKIISGLRDNSIKIWDKTSLECLKVLTGHTGSVLCLQYDERVIVTGSSDSTVRVWDVNTGEVLNTLIHHNEAVLHLRFSNGLMVTCSKDRSIAVWDMASATDITLRRVLVGHRAAVNVVDFDDKYIVSASGDRTIKVWSTSTCEFVRTLNGHKRGIACLQYRDRLVVSGSSDNTIRLWDIECGACLRVLEGHEELVRCIRFDNKRIVSGAYDGKIKVWDLQAALDPRAPASTLCLRTLVEHSGRVFRLQFDEFQIISSSHDDTILIWDFLNVPPSAQNETRSPSRTYTYISR

Specifications

Antigen F-box and WD repeat domain containing 11
Applications ELISA, Immunofluorescence, In situ PLA
Classification Monoclonal
Clone 2E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant FBXW11.
Formulation PBS with no preservative; pH 7.4
Gene FBXW11
Gene Accession No. BC026213
Gene Alias BTRC2/BTRCP2/FBW1B/FBXW1B/Fbw11/Hos/KIAA0696
Gene Symbols FBXW11
Host Species Mouse
Immunogen FBXW11 (AAH26213, 1 a.a. ∼ 529 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23291
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.