Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523773
Description
Members of the F-box protein family, such as FBXW12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603034), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications
| FBXW12 | |
| Polyclonal | |
| FBXW12 | |
| FBW12, FBXO12, FBXO35, F-box and WD-40 domain protein 12, F-box and WD-40 domain-containing protein 12, F-box only protein 35, F-box- and WD40-repeat-containing protein, F-box/WD repeat-containing protein 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285231 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q6X9E4 | |
| FBXW12 | |
| KLH conjugated synthetic peptide between 291-320 amino acids from the C-terminal region of human FBXW12 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction