Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box and WD repeat domain containing 5, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-095 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-095 Supplier Abnova Corporation Supplier No. H00054461B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FBXW5 protein.

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains WD-40 domains, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene, however, they were found to be nonsense-mediated mRNA decay (NMD) candidates, hence not represented. [provided by RefSeq]

Sequence: MGLSPDNRYLYVNSRAWPNGAVVADPMQPPPIAEEIDLLVFDLKTMREVRRALRAHRAYTPNDECFFIFLDVSRDFVASGAEDRHGYIWDRHYNICLARLRHEDVVNSVVFSPQEQELLLTASDDATIKAWRSPRTMRVLQAPRPRPRTFFSWLASQRR

Specifications

Antigen F-box and WD repeat domain containing 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FBXW5 protein.
Formulation PBS with no preservative; pH 7.4
Gene FBXW5
Gene Accession No. BC000850.1
Gene Alias DKFZp434B205/Fbw5/MGC20962/RP11-229P13.10
Gene Symbols FBXW5
Host Species Mouse
Immunogen FBXW5 (AAH00850.1, 1 a.a. ∼ 159 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Primary or Secondary Primary
Gene ID (Entrez) 54461
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.