Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fcμ Receptor Mouse anti-Human, Alexa Fluor 647, Clone: HM14-1 (also known as HM14), BD

Catalog No. BDB563535 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563535 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563535 Supplier BD Biosciences Supplier No. 563535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HM14-1 monoclonal antibody specifically binds to the FAIM3-encoded high affinity receptor for IgM (also known as, Fcµ Receptor, FcλR, Fc mu receptor, Immunoglobulin mu Fc receptor, FcmR, IgM Fc receptor). The FcλR is a type 1 transmembrane glycoprotein that contains an extracellular Ig-like domain with sequence similarities to Fcα/λR and pIgR. It is likewise known as Fas apoptotic inhibitory molecule 3, TOSO or Regulator of Fas-induced apoptosis Toso based on its capacity to counter apoptosis mediated by Fas/CD95 (or TNF) in some experimental model systems. FcλRs are expressed by B cells, CD4+ T cells, CD8+ T cells, and NK cells and are highly expressed on Chronic Lymphocytic Leukemia cells. FcλRs may play roles in cellular activation, antigen presentation and IgM-dependent cell-mediated cytotoxicity. The relatively long cytoplasmic tail of FcλR contains several conserved tyrosine and serine residues. These get phosphorylated after FcλR ligation with IgM immune complexes and may initiate signaling responses. Lymphocyte FcλRscan mediate the internalization of IgM-bound antigens including microbial and viral antigens. This in turn may cause cellular activation due to subsequent antigen processing and exposure of activating ligands to intracellular receptors including Toll-like receptors. The HM7-10 antibody, a related FcμR-specific antibody, reportedly recognizes an epitope near the IgM ligand binding site of the FcμR whereas the HM14-1 antibody does not. oll-like receptors. The HM7-10 antibody, a related FcλR-specific antibody, reportedly recognizes an epitope near the IgM ligand binding site of the FcλR whereas the HM14-1 antibody does not.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fcμ Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone HM14-1 (also known as HM14)
Conjugate Alexa Fluor 647
Description FcμR; FcmR; Fc mu receptor; IgM Fc receptor; FAIM3; TOSO
Host Species Mouse
Immunogen Human IgM Fc Receptor Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.