Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fcμ Receptor Mouse anti-Human, PE, Clone: HM7-10, BD

Catalog No. BDB563019 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563019 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563019 Supplier BD Biosciences Supplier No. 563019

Mouse Monoclonal Antibody

The HM7-10 monoclonal antibody specifically binds to the FAIM3 -encoded high affinity receptor for IgM (also known as, Fcµ Receptor, FcµR, Fc mu receptor, Immunoglobulin mu Fc receptor, FcmR, IgM Fc receptor). The FcµR is a type 1 transmembrane glycoprotein that contains an extracellular Ig-like domain with sequence similarities to Fc α /µR and pIgR. It is likewise known as Fas apoptotic inhibitory molecule 3, TOSO or Regulator of Fas-induced apoptosis Toso based on its capacity to counter apoptosis mediated by Fas/CD95 (or TNF) in some experimental model systems. FcµRs are expressed by B cells, CD4+ T cells, CD8+ T cells, and NK cells and are highly expressed on Chronic Lymphocytic Leukemia cells. FcµRs may play roles in cellular activation, antigen presentation and IgM-dependent cell-mediated cytotoxicity. The relatively long cytoplasmic tail of FcµR contains several conserved tyrosine and serine residues. These get phosphorylated after FcµR ligation with IgM immune complexes and may initiate signaling responses. Lymphocyte FcµRs can mediate the internalization of IgM-bound antigens including microbial and viral antigens. This in turn may cause cellular activation due to subsequent antigen processing and exposure of activating ligands to intracellular receptors including Toll-like receptors. The HM7-10 antibody reportedly recognizes an epitope near the IgM ligand binding site of the FcµR whereas, a related FcµR-specific antibody, HM14-1 antibody does not. The HM7 epitope is sensitive to IgM ligand binding.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fcμ Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone HM7-10
Conjugate PE
Description FcμR; FcmR; Fc mu receptor; IgM Fc receptor; FAIM3; TOSO
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IgM Fc Receptor Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.