Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Fc Receptor I γ subunit; Polyclonal; ™ (Upstate™)
Supplier: MilliporeSigma™ 06727/DEL

Description
Repetto et. al., J. Immunol. 15:4876, 1996
Immunoprecipitation, Western Blotting

Specifications
Fc Receptor I γ subunit | |
Unconjugated | |
Peptide KHEKPPQ corresponding to amino acids 80-86 of γ-subunit of Fc receptors | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
200 μg | |
Inflammation, Immunology | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction