Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcΕRI α subunit Mouse anti-Rat, MilliporeSigma™

Catalog No. 05468MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-468-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-468-MI Supplier MilliporeSigma Supplier No. 05468
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FcΕRI α subunit Antibody is an antibody against FcΕRI α subunit for use in FC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcεRI α subunit
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, 0.15M NaCl, 0.05% sodium azide prior to the addition of glycerol to 30%
Gene Accession No. NM_002001
Gene Symbols FCER1A, FcERI, FCE1A
Host Species Mouse
Immunogen Whole rat basophilic leukemia cells, positive for the rat high-affinity receptor recognizing immunoglobulin E (FcεRI).
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.