Learn More
FcΕRI Rabbit anti-Rat, Human, Mouse, FITC, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | FcεRI |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA. |
| Gene Symbols | FCER1G; OTTHUMP00000063478 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids 80-86 of γ-subunit of Fc receptors. |
| Purification Method | Protein A Purified |
| Quantity | 100 Tests |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.